7-(Trifluormethyl)-4-chinolinol

Lieferant: FLUOROCHEM

Synonyme: 4-Hydroxy-7-(trifluoromethyl)quinoline

CFF1183.1 007674-1G
CHMPCFF1183.1EA 522 EUR
CHMPCFF1183.1 FLUO007674-1G
7-(Trifluormethyl)-4-chinolinol
7-(Trifluormethyl)-4-chinolinol
Formel: C₁₀H₆F₃NO
Molecular Weight: 213,1560 g/mol
MDL: MFCD00006779
CAS-Nummer: 322-97-4
EINECS: 206-298-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR