Trifluor(trifluormethoxy)ethylen
Lieferant: FLUOROCHEM
007684-100G
FLUO007684-100GEA
477
EUR
FLUO007684-100G
Trifluor(trifluormethoxy)ethylen
Trifluor(trifluormethoxy)ethylen
Formel:
C₃F₆O Siedepunkt: –22 °C (1013 hPa) Schmelzpunkt: –125 °C |
MDL:
MFCD00221780 CAS-Nummer: 1187-93-5 EINECS: 214-703-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...