2,4,6-Trifluorpyrimidin

Lieferant: FLUOROCHEM
Gefahr

007722-1G
FLUO007722-1GEA 69.9 EUR
FLUO007722-1G
2,4,6-Trifluorpyrimidin
2,4,6-Trifluoropyrimidine
Formel: C₄HF₃N₂
Molecular Weight: 134,06 g/mol
Siedepunkt: 59…60 °C (1013 hPa)
MDL: MFCD00039705
CAS-Nummer: 696-82-2
EINECS: 211-801-1
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR