Yttrium(III)fluorid
Lieferant: FLUOROCHEM
Synonyme:
Yttriumtrifluorid
007781-25G
007781-100G
FLUO007781-25GEA
51.8
EUR
FLUO007781-25G
FLUO007781-100G
Yttrium(III)fluorid
Yttrium(III)fluorid
Formel:
F₃Y Molecular Weight: 145.9 g/mol Siedepunkt: 2227 Schmelzpunkt: 1387 Dichte: 4.01 g/cm³ (20 °C) |
MDL:
MFCD00049714 CAS-Nummer: 13709-49-4 EINECS: 237-257-5 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...