3-Brom-4-(trifluormethoxy)anilin

Lieferant: FLUOROCHEM

007803-5G 007803-1G 007803-25G
FLUO007803-5GEA 36.2 EUR
FLUO007803-5G FLUO007803-1G FLUO007803-25G
3-Brom-4-(trifluormethoxy)anilin
3-Brom-4-(trifluormethoxy)anilin
Formel: C₇H₅BrF₃NO
MDL: MFCD00190127
CAS-Nummer: 191602-54-7
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR