4-Brom-2,6-difluorphenol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1-Bromo-3,5-difluoro-4-hydroxybenzene

007833-5G
FLUO007833-5GEA 42.9 EUR
FLUO007833-5G
4-Brom-2,6-difluorphenol
4-Brom-2,6-difluorphenol
Formel: C₆H₃BrF₂O
MDL: MFCD00236213
CAS-Nummer: 104197-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR