3-(Difluormethoxy)benzonitril

Lieferant: FLUOROCHEM

007854-1G
FLUO007854-1GEA 69.8 EUR
FLUO007854-1G
3-(Difluormethoxy)benzonitril
3-(Difluormethoxy)benzonitril
Formel: C₈H₅F₂NO
Molecular Weight: 169,1282 g/mol
MDL: MFCD00236225
CAS-Nummer: 97582-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR