3-(Difluormethoxy)benzylamin

Lieferant: FLUOROCHEM

007859-1G
FLUO007859-1GEA 152 EUR
FLUO007859-1G
3-(Difluormethoxy)benzylamin
3-(Difluormethoxy)benzylamin
Formel: C₈H₉F₂NO
MDL: MFCD00236230
CAS-Nummer: 244022-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR