3-(Difluormethoxy)phenylacetonitril
Lieferant: FLUOROCHEM
007871-1G
007871-2.5G
FLUO007871-1GEA
488
EUR
FLUO007871-1G
FLUO007871-2.5G
3-(Difluormethoxy)phenylacetonitril
3-(Difluormethoxy)phenylacetonitril
Formel:
C₉H₇F₂NO |
MDL:
MFCD00236241 CAS-Nummer: 41429-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...