3'-Fluor-4'-(trifluormethyl)acetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Fluoro-4-(trifluoromethyl)phenyl)ethanone

007905-5G 007905-1G
FLUO007905-5GEA 231 EUR
FLUO007905-5G FLUO007905-1G
3'-Fluor-4'-(trifluormethyl)acetophenon
3'-Fluor-4'-(trifluormethyl)acetophenon
Formel: C₉H₆F₄O
MDL: MFCD00236273
CAS-Nummer: 237761-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR