5-Fluor-2-(trifluormethyl)benzamid

Lieferant: FLUOROCHEM

007910-5G 007910-1G
FLUO007910-5GEA 222 EUR
FLUO007910-5G FLUO007910-1G
5-Fluor-2-(trifluormethyl)benzamid
5-Fluor-2-(trifluormethyl)benzamid
Formel: C₈H₅F₄NO
CAS-Nummer: 654-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR