2-Fluor-3-(trifluormethyl)zimtsäure

Lieferant: FLUOROCHEM

007926-1G 007926-5G
FLUO007926-1GEA 39.9 EUR
FLUO007926-1G FLUO007926-5G
2-Fluor-3-(trifluormethyl)zimtsäure
2-Fluor-3-(trifluormethyl)zimtsäure
Formel: C₁₀H₆F₄O₂
MDL: MFCD00236291
CAS-Nummer: 237069-83-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR