2-Fluor-4-(trifluormethyl)zimtsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(2-Fluoro-4-(trifluoromethyl)phenyl)acrylic acid

007927-1G 007927-5G
FLUO007927-1GEA 0 EUR
FLUO007927-1G FLUO007927-5G
2-Fluor-4-(trifluormethyl)zimtsäure
2-Fluor-4-(trifluormethyl)zimtsäure
Formel: C₁₀H₆F₄O₂
MDL: MFCD00236292
CAS-Nummer: 262608-88-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR