4-Fluor-2-(trifluormethyl)zimtsäure

Lieferant: FLUOROCHEM

007932-5G 007932-1G
FLUO007932-5GEA 177 EUR
FLUO007932-5G FLUO007932-1G
4-Fluor-2-(trifluormethyl)zimtsäure
4-Fluor-2-(trifluormethyl)zimtsäure
Formel: C₁₀H₆F₄O₂
MDL: MFCD00236297
CAS-Nummer: 243977-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR