α-Chlor-2,4,5-trifluortoluol

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 2,4,5-Trifluorbenzylchlorid, 1-(Chlormethyl)-2,4,5-trifluorbenzol

007969-1G 007969-5G 007969-10G
FLUO007969-1GEA 34.1 EUR
FLUO007969-1G FLUO007969-5G FLUO007969-10G
α-Chlor-2,4,5-trifluortoluol
α-Chlor-2,4,5-trifluortoluol
Formel: C₇H₄ClF₃
Molecular Weight: 180.56 g/mol
MDL: MFCD00236319
CAS-Nummer: 243139-71-1
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR