2'-Trifluormethoxyacetophenon

Lieferant: CHEMPUR

Synonyme: 1-[2-(Trifluormethoxy)phenyl]-1-ethanon

FL7976.POR 007976-1G 007976-5G 007976-25G
CHMPFL7976.POREA 0 EUR
CHMPFL7976.POR FLUO007976-1G FLUO007976-5G FLUO007976-25G
2'-Trifluormethoxyacetophenon
2'-Trifluormethoxyacetophenon
Formel: C₉H₇F₃O₂
Molecular Weight: 204.15
MDL: MFCD00236320
CAS-Nummer: 220227-93-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR