3-((Trifluormethyl)thio)benzoesäure

Lieferant: FLUOROCHEM

007998.25G 007998-5G 007998-1G
FLUO007998.25GEA 516 EUR
FLUO007998.25G FLUO007998-5G FLUO007998-1G
3-((Trifluormethyl)thio)benzoesäure
3-((Trifluormethyl)thio)benzoesäure
Formel: C₈H₅F₃O₂S
MDL: MFCD00236337
CAS-Nummer: 946-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR