4-(Trifluormethylthio)benzylalkohol
Lieferant: FLUOROCHEM
008006-1G
FLUO008006-1GEA
108
EUR
FLUO008006-1G
4-(Trifluormethylthio)benzylalkohol
4-(Trifluormethylthio)benzylalkohol
Formel:
C₈H₇F₃OS |
MDL:
MFCD00236342 CAS-Nummer: 56456-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...