3-(Trifluormethylthio)phenylessigsäure

Lieferant: FLUOROCHEM

008015-1G
FLUO008015-1GEA 121 EUR
FLUO008015-1G
3-(Trifluormethylthio)phenylessigsäure
3-(Trifluormethylthio)phenylessigsäure
Formel: C₉H₇F₃O₂S
Molecular Weight: 236,21 g/mol
MDL: MFCD00236350
CAS-Nummer: 239080-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR