5-Brom-m-xylol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
1-Brom-3,5-dimethylbenzol, 3,5-Dimethylbrombenzol
008025-25G
008025-10G
008025-50G
FLUO008025-25GEA
60.9
EUR
FLUO008025-25G
FLUO008025-10G
FLUO008025-50G
5-Brom-m-xylol
5-Brom-m-xylol
Formel:
C₈H₉Br Molecular Weight: 185.06 g/mol Siedepunkt: 60…62 °C (5.33 hPa) Dichte: 1.36 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000087 CAS-Nummer: 556-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...