(Isopentyl)triphenylphosphoniumbromid

Lieferant: FLUOROCHEM
Achtung

Synonyme: (Isoamyl)triphenylphosphoniumbromid

008045-25G 008045-100G
FLUO008045-25GEA 0 EUR
FLUO008045-25G FLUO008045-100G
(Isopentyl)triphenylphosphoniumbromid
(Isopentyl)triphenylphosphoniumbromid
Formel: (H₃C)₂HCCH₂CH₂P(C₆H₅)₃Br
Molecular Weight: 413.34 g/mol
MDL: MFCD00031594
CAS-Nummer: 28322-40-9
EINECS: 248-966-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR