1-(Methylsulfonyl)-4-(trifluormethoxy)benzol

Lieferant: FLUOROCHEM

008060-1G 008060-5G
FLUO008060-1GEA 45.9 EUR
FLUO008060-1G FLUO008060-5G
1-(Methylsulfonyl)-4-(trifluormethoxy)benzol
1-(Methylsulphonyl)-4-(trifluoromethoxy)benzene
Formel: C₈H₇F₃O₃S
MDL: MFCD00792426
CAS-Nummer: 87750-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR