Methylthiosalicylat

Lieferant: FLUOROCHEM

Synonyme: Methyl 2-mercaptobenzoate

008066-5G 008066-25G
FLUO008066-5GEA 120 EUR
FLUO008066-5G FLUO008066-25G
Methylthiosalicylat
Methylthiosalicylat
Formel: C₈H₈O₂S
MDL: MFCD00060678
CAS-Nummer: 4892-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR