3-Nitrophenylsulfurpentafluorid

Lieferant: FLUOROCHEM

CFF1244. 008099-1G 008099-5G
CHMPCFF1244.EA 0 EUR
CHMPCFF1244. FLUO008099-1G FLUO008099-5G
3-Nitrophenylsulfurpentafluorid
3-Nitrophenylsulfurpentafluorid
Formel: C₆H₄F₅NO₂S
Molecular Weight: 249.16 g/mol
MDL: MFCD00221621
CAS-Nummer: 2613-26-5
EINECS: 447-600-2
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR