α,α-Difluoranisol

Lieferant: FLUOROCHEM

Synonyme: (Difluoromethoxy)benzene, alpha,alpha-Difluoroanisole

008159-100G
FLUO008159-100GEA 317 EUR
FLUO008159-100G
α,α-Difluoranisol
α,α-Difluoranisol
Formel: C₇H₆F₂O
Molecular Weight: 144,1200 g/mol
MDL: MFCD00068184
CAS-Nummer: 458-92-4
EINECS: 207-283-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR