3,4-Difluor-o-phenylendiamin ≥98%

Lieferant: FLUOROCHEM

Synonyme: 3,4-Difluorbenzol-1,2-diamin, 3,4-Difluor-1,2-phenylendiamin, 1,2-Diamino-3,4-difluorbenzol

008166-1G
FLUO008166-1GEA 69.6 EUR
FLUO008166-1G
3,4-Difluor-o-phenylendiamin ≥98%
3,4-Difluor-o-phenylendiamin
Formel: C₆H₆F₂N₂
Molecular Weight: 144.12 g/mol
MDL: MFCD00153128
CAS-Nummer: 153505-39-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR