N-(2,4-Difluorphenyl)-2-chloracetamid

Lieferant: FLUOROCHEM

Synonyme: 2-Chloro-2',4'-difluoroacetanilide, 2-Chloro-N-(2,4-difluorophenyl)acetamide

008219-5G
FLUO008219-5GEA 142 EUR
FLUO008219-5G
N-(2,4-Difluorphenyl)-2-chloracetamid
N-(2,4-Difluorphenyl)-2-chloracetamid
Formel: C₈H₆ClF₂NO
Molecular Weight: 205,5892 g/mol
MDL: MFCD00077473
CAS-Nummer: 96980-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR