4-Chlor-6-fluor-2-(trifluormethyl)chinolin

Lieferant: FLUOROCHEM

008244-1G
FLUO008244-1GEA 296 EUR
FLUO008244-1G
4-Chlor-6-fluor-2-(trifluormethyl)chinolin
4-Chloro-6-fluoro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₄ClF₄N
MDL: MFCD00276579
CAS-Nummer: 59611-55-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR