3-(Chlormethyl)-5-(4-(trifluormethyl)phenyl)-1,2,4-oxadiazol

Lieferant: FLUOROCHEM

008253-1G
FLUO008253-1GEA 395 EUR
FLUO008253-1G
3-(Chlormethyl)-5-(4-(trifluormethyl)phenyl)-1,2,4-oxadiazol
3-(Chloromethyl)-5-(4-(trifluoromethyl)phenyl)-1,2,4-oxadiazole
Formel: C₁₀H₆ClF₃N₂O
MDL: MFCD00173952
CAS-Nummer: 175205-84-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR