1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)piperazin

Lieferant: FLUOROCHEM

008265-1G
FLUO008265-1GEA 76.2 EUR
FLUO008265-1G
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)piperazin
1-(3-Chlor-5-(trifluormethyl)-2-pyridinyl)piperazin
Formel: C₁₀H₁₁ClF₃N₃
MDL: MFCD00173934
CAS-Nummer: 132834-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR