2-(2,4-Difluorphenoxy)-3-nitropyridin
Lieferant: FLUOROCHEM
008287-1G
FLUO008287-1GEA
87.9
EUR
FLUO008287-1G
2-(2,4-Difluorphenoxy)-3-nitropyridin
2-(2,4-Difluorophenoxy)-3-nitropyridine
Formel:
C₁₁H₆F₂N₂O₃ |
MDL:
MFCD00067800 CAS-Nummer: 175135-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...