N-(2,4-Difluorphenyl)maleamidsäure
Lieferant: FLUOROCHEM
008290-1G
FLUO008290-1GEA
104
EUR
FLUO008290-1G
N-(2,4-Difluorphenyl)maleamidsäure
N-(2,4-Difluorphenyl)maleamidsäure
Formel:
C₁₀H₇F₂NO₃ |
MDL:
MFCD00082911 CAS-Nummer: 6954-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...