Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate

008297-5G 008297-1G
FLUO008297-5GEA 155 EUR
FLUO008297-5G FLUO008297-1G
Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-(4-fluorphenyl)-1H-pyrazol-4-carboxylat
Formel: C₁₂H₁₂FN₃O₂
MDL: MFCD00173917
CAS-Nummer: 138907-68-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR