3-Fluor-4-methylbenzamidoxim

Lieferant: FLUOROCHEM

008340-1G
FLUO008340-1GEA 116 EUR
FLUO008340-1G
3-Fluor-4-methylbenzamidoxim
3-Fluor-4-methylbenzamidoxim
Formel: C₈H₉FN₂O
MDL: MFCD00673178
CAS-Nummer: 175277-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR