2-Fluor-6-(4-(methylthio)phenoxy)benzonitril

Lieferant: FLUOROCHEM

008350-1G
FLUO008350-1GEA 94.1 EUR
FLUO008350-1G
2-Fluor-6-(4-(methylthio)phenoxy)benzonitril
2-Fluor-6-(4-(methylthio)phenoxy)benzonitril
Formel: C₁₄H₁₀FNOS
MDL: MFCD00068069
CAS-Nummer: 148901-52-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR