(3-Trifluormethyl-α-toluolsulfonyl)acetonitril

Lieferant: FLUOROCHEM

008435-1G
FLUO008435-1GEA 128 EUR
FLUO008435-1G
(3-Trifluormethyl-α-toluolsulfonyl)acetonitril
(3-Trifluormethyl-α-toluolsulfonyl)acetonitril
Formel: C₁₀H₈F₃NO₂S
MDL: MFCD00173822
CAS-Nummer: 175276-81-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR