2-Brom-1-ethylpyridiniumtetrafluoroborat

Lieferant: FLUOROCHEM

Synonyme: 2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate

008541-5G
FLUO008541-5GEA 213 EUR
FLUO008541-5G
2-Brom-1-ethylpyridiniumtetrafluoroborat
2-Bromo-1-ethylpyridinium tetrafluoroborate
Formel: C₇H₉BBrF₄N
Molecular Weight: 273,86 g/mol
MDL: MFCD00060046
CAS-Nummer: 878-23-9
EINECS: 212-900-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR