4,4'-Difluorbenzil

Lieferant: FLUOROCHEM
Achtung

Synonyme: Bis(4-fluorphenyl)ethandion

008552-5G
FLUO008552-5GEA 104 EUR
FLUO008552-5G
4,4'-Difluorbenzil
4,4'-Difluorbenzil
Formel: C₁₄H₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00134541
CAS-Nummer: 579-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR