3-Fluor-L-phenylalanin

Lieferant: FLUOROCHEM

008570-100MG
FLUO008570-100MGEA 0 EUR
FLUO008570-100MG
3-Fluor-L-phenylalanin
3-Fluor-L-phenylalanin
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,1796 g/mol
MDL: MFCD00066450
CAS-Nummer: 19883-77-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR