2,2,2-Trifluorethylmethansulfonat

Lieferant: FLUOROCHEM

008580-5G
FLUO008580-5GEA 0 EUR
FLUO008580-5G
2,2,2-Trifluorethylmethansulfonat
2,2,2-Trifluorethylmethansulfonat
Formel: C₃H₅F₃O₃S
Molecular Weight: 178,130 g/mol
MDL: MFCD00025058
CAS-Nummer: 25236-64-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR