4-Fluor-7-nitrobenzofurazan

Lieferant: FLUOROCHEM

Synonyme: 4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole, NBD-F

008587-100MG
FLUO008587-100MGEA 0 EUR
FLUO008587-100MG
4-Fluor-7-nitrobenzofurazan
4-Fluor-7-nitrobenzofurazan
Formel: C₆H₂FN₃O₃
Molecular Weight: 183,1 g/mol
MDL: MFCD00010196
CAS-Nummer: 29270-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR