1,1,1,2,3,3-Hexafluor-2-(heptafluorpropoxy)-3-[(trifluorvinyl)oxy]propan
Lieferant: FLUOROCHEM
008603-5G
FLUO008603-5GEA
170
EUR
FLUO008603-5G
1,1,1,2,3,3-Hexafluor-2-(heptafluorpropoxy)-3-[(trifluorvinyl)oxy]propan
1,1,1,2,3,3-Hexafluor-2-(heptafluorpropoxy)-3-[(trifluorvinyl)oxy]propan
Formel:
F₂CCFOCF₂CF(CF₃)O(CF₂)₂CF₃ |
MDL:
MFCD00191479 CAS-Nummer: 1644-11-7 EINECS: 216-703-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...