Tetrabutylammoniumdihydrogentrifluorid
Lieferant: FLUOROCHEM
008617-5G
FLUO008617-5GEA
72.9
EUR
FLUO008617-5G
Tetrabutylammoniumdihydrogentrifluorid
Tetrabutylammoniumdihydrogentrifluorid
Formel:
C₁₆H₃₈F₃N |
MDL:
MFCD00145365 CAS-Nummer: 99337-56-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...