2,3,5,6-Tetrafluor-4-(trifluormethyl)anilin

Lieferant: FLUOROCHEM

Synonyme: 4-Aminotetrafluorobenzotrifluoride

008618-5G 008618-1G
FLUO008618-5GEA 267 EUR
FLUO008618-5G FLUO008618-1G
2,3,5,6-Tetrafluor-4-(trifluormethyl)anilin
2,3,5,6-Tetrafluor-4-(trifluormethyl)anilin
Formel: C₇H₂F₇N
Molecular Weight: 233.09 g/mol
MDL: MFCD00091518
CAS-Nummer: 651-83-2
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR