4-(Trifluormethyl)benzoesäureanhydrid

Lieferant: FLUOROCHEM

008630-10G
FLUO008630-10GEA 377 EUR
FLUO008630-10G
4-(Trifluormethyl)benzoesäureanhydrid
4-(Trifluoromethyl)benzoic anhydride
Formel: C₁₆H₈F₆O₃
Molecular Weight: 362,23 g/mol
MDL: MFCD00671577
CAS-Nummer: 25753-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR