1-Chlor-2,4-dibrom-6-fluorbenzol
Lieferant: FLUOROCHEM
008670-5G
FLUO008670-5GEA
73.2
EUR
FLUO008670-5G
1-Chlor-2,4-dibrom-6-fluorbenzol
1-Chloro-2,4-dibromo-6-fluorobenzene
Formel:
C₆H₂Br₂ClF |
MDL:
MFCD00142891 CAS-Nummer: 202925-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...