1,2-Dibrom-4-fluorbenzol

Lieferant: FLUOROCHEM

008673-5G
FLUO008673-5GEA 0 EUR
FLUO008673-5G
1,2-Dibrom-4-fluorbenzol
1,2-Dibromo-4-fluorobenzene
Formel: C₆H₃Br₂F
MDL: MFCD00070780
CAS-Nummer: 2369-37-1
EINECS: 219-131-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR