3-Chlor-4-methylphenylisothiocyanat
Lieferant: FLUOROCHEM
008766-5G
FLUO008766-5GEA
69.9
EUR
FLUO008766-5G
3-Chlor-4-methylphenylisothiocyanat
3-Chlor-4-methylphenylisothiocyanat
Formel:
C₈H₆ClNS Siedepunkt: 263 °C (1013 hPa) Dichte: 1,24 g/cm³ (20 °C) |
MDL:
MFCD00022057 CAS-Nummer: 19241-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...