2-Chlor-5-(trifluormethyl)phenylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1-Chloro-2-isothiocyanato-4-(trifluoromethyl)benzene
008776-5G
FLUO008776-5GEA
63.9
EUR
FLUO008776-5G
2-Chlor-5-(trifluormethyl)phenylisothiocyanat
2-Chlor-5-(trifluormethyl)phenylisothiocyanat
Formel:
C₈H₃ClF₃NS Molecular Weight: 237.63 g/mol Siedepunkt: 114…117 °C (1013 hPa) Dichte: 1.45 g/cm³ (20 °C) |
MDL:
MFCD00041055 CAS-Nummer: 23165-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...