1-Fluor-3-ethinylbenzol
Lieferant: FLUOROCHEM
Synonyme:
1-Ethinyl-3-fluorbenzol, (3-Fluorphenyl)acetylen, (3-Fluorphenyl)ethin
008807-1G
FLUO008807-1GEA
45.9
EUR
FLUO008807-1G
1-Fluor-3-ethinylbenzol
1-Fluor-3-ethinylbenzol
Formel:
C₈H₅F Molecular Weight: 120.13 g/mol |
MDL:
MFCD03093798 CAS-Nummer: 2561-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...